| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR1300004443.mm.1 |
NM_001282055.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300004443.mm.1 |
TC1300000671.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1300004445.mm.1 |
NM_001282055.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300004445.mm.1 |
TC1300000671.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1300004441.mm.1 |
NM_001282055.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300004441.mm.1 |
TC1300000671.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1300004444.mm.1 |
NM_001282055.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300004444.mm.1 |
TC1300000671.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1300004446.mm.1 |
NM_001282055.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300004446.mm.1 |
TC1300000671.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1300004442.mm.1 |
NM_001282055.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300004442.mm.1 |
TC1300000671.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1300004439.mm.1 |
NM_001282055.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300004439.mm.1 |
TC1300000671.mm.1 |
MTA TC |