Assay Details
| Gene Symbol: Ddx46 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001282055.1 | NP_001268984.1 | 6 - 7 | 952 | 86 | |
| GenBank mRNA | AK088950.1 | - | 6 - 7 | 924 | 86 | |
| AK129220.1 | - | 6 - 7 | 925 | 86 | ||
| BC092240.1 | - | 5 - 6 | 863 | 30662573 | 86 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ddx46
Genomic Map