Assay Details
| Gene Symbol: Adamtsl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_029981.1 | NP_084257.1 | 15 - 16 | 2455 | 85 | |
| GenBank mRNA | AB096928.1 | - | 15 - 16 | 2455 | 85 | |
| AK172978.1 | - | 3 - 4 | 4861 | 85 | ||
| BC128568.1 | - | 15 - 16 | 2407 | 40097415 | 85 | |
| BC129984.1 | - | 15 - 16 | 2407 | 40097418 | 85 | |
More Information
Updated Mapping Information
Set Membership: |
|

Adamtsl2
Genomic Map