Assay Details
| Gene Symbol: Adamtsl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_029981.1 | NP_084257.1 | 17 - 18 | 2799 | 74 | |
| GenBank mRNA | AB096928.1 | - | 17 - 18 | 2799 | 74 | |
| AK020831.1 | - | 1 - 2 | 261 | 74 | ||
| AK156991.1 | - | 17 - 18 | 2802 | 74 | ||
| AK172978.1 | - | 5 - 6 | 5205 | 74 | ||
| BC128568.1 | - | 17 - 18 | 2751 | 40097415 | 74 | |
| BC129984.1 | - | 17 - 18 | 2751 | 40097418 | 74 | |
More Information
Updated Mapping Information
Set Membership: |
|

Adamtsl2
Genomic Map