Assay Details
| Gene Symbol: Adamtsl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_029981.1 | NP_084257.1 | 6 - 7 | 748 | 100 | |
| GenBank mRNA | AB096928.1 | - | 6 - 7 | 748 | 100 | |
| AK156991.1 | - | 6 - 7 | 751 | 100 | ||
| BC128568.1 | - | 6 - 7 | 700 | 40097415 | 100 | |
| BC129984.1 | - | 6 - 7 | 700 | 40097418 | 100 | |
More Information
Updated Mapping Information
Set Membership: |
|

Adamtsl2
Genomic Map