| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0200027860.mm.1 |
NM_001038700.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200027860.mm.1 |
NM_001038700.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200027860.mm.1 |
NM_001177648.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200027860.mm.1 |
NM_001177648.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200027860.mm.1 |
NM_001177649.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200027860.mm.1 |
NM_001177649.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200027860.mm.1 |
NM_001177650.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200027860.mm.1 |
NM_001177650.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200027860.mm.1 |
TC0200003283.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0200027865.mm.1 |
NM_001177648.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200027865.mm.1 |
NM_001177648.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200027865.mm.1 |
NM_001177650.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200027865.mm.1 |
NM_001177650.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200027865.mm.1 |
TC0200003283.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0200027861.mm.1 |
NM_001038700.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200027861.mm.1 |
NM_001038700.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200027861.mm.1 |
NM_001177648.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200027861.mm.1 |
NM_001177648.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200027861.mm.1 |
NM_001177649.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200027861.mm.1 |
NM_001177649.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200027861.mm.1 |
NM_001177650.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200027861.mm.1 |
NM_001177650.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200027861.mm.1 |
TC0200003283.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0200027864.mm.1 |
NM_001177648.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200027864.mm.1 |
NM_001177648.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200027864.mm.1 |
NM_001177650.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200027864.mm.1 |
NM_001177650.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200027864.mm.1 |
TC0200003283.mm.1 |
MTA TC |