Assay Details
| Gene Symbol: Unc5a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_153131.3 | NP_694771.1 | 10 - 11 | 1860 | 65 | |
| GenBank mRNA | AJ487852.1 | - | 10 - 11 | 1861 | 65 | |
| BC058084.1 | - | 9 - 10 | 1713 | 6813463 | 65 | |
More Information
Updated Mapping Information
Set Membership: |
|

Unc5a
Genomic Map