Assay Details
| Gene Symbol: Unc5a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_153131.3 | NP_694771.1 | 9 - 10 | 1775 | 122 | |
| GenBank mRNA | AJ487852.1 | - | 9 - 10 | 1776 | 122 | |
| BC058084.1 | - | 8 - 9 | 1628 | 6813463 | 122 | |
More Information
Updated Mapping Information
Set Membership: |
|

Unc5a
Genomic Map