Assay Details
| Gene Symbol: Psmb7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011187.1 | NP_035317.1 | 2 - 3 | 153 | 70 | |
| GenBank mRNA | AK002823.1 | - | 2 - 3 | 168 | 70 | |
| AK012613.1 | - | 2 - 3 | 154 | 70 | ||
| AK013961.1 | - | 2 - 3 | 164 | 70 | ||
| AK031913.1 | - | 2 - 3 | 169 | 70 | ||
| AK075759.1 | - | 2 - 3 | 169 | 70 | ||
| AK088276.1 | - | 2 - 3 | 164 | 70 | ||
| AK088765.1 | - | 2 - 3 | 167 | 70 | ||
| AK162002.1 | - | 2 - 3 | 172 | 70 | ||
| AK168823.1 | - | 2 - 3 | 156 | 70 | ||
| BC033424.1 | - | 2 - 3 | 150 | 70 | ||
| BC049230.1 | - | 2 - 3 | 146 | 70 | ||
| BC057662.1 | - | 2 - 3 | 154 | 2865594 | 70 | |
| D83585.1 | - | 2 - 3 | 153 | 70 | ||
| Y10874.1 | - | 2 - 3 | 167 | 70 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Psmb7
Genomic Map