Assay Details
| Gene Symbol: Ptpdc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001301781.1 | NP_001288710.1 | 8 - 9 | 2326 | 108 | |
| NM_207232.2 | NP_997115.1 | 7 - 8 | 2143 | 108 | ||
| GenBank mRNA | AK030415.1 | - | 7 - 8 | 2136 | 108 | |
| AK038922.1 | - | 7 - 8 | 2056 | 108 | ||
| AK132781.1 | - | 8 - 9 | 2235 | 108 | ||
| BC066081.1 | - | 7 - 8 | 2044 | 30355705 | 108 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ptpdc1
Genomic Map