Assay Details
| Gene Symbol: Fam120a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001033268.2 | NP_001028440.2 | 10 - 11 | 1900 | 92 | |
| GenBank mRNA | AK172909.1 | - | 10 - 11 | 2089 | 92 | |
| BC010304.1 | - | 6 - 7 | 968 | 92 | ||
| BC033412.1 | - | 2 - 3 | 228 | 92 | ||
| BC042582.1 | - | 9 - 10 | 1416 | 92 | ||
| BC158134.1 | - | 10 - 11 | 1892 | 92 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Fam120a
Genomic Map