Assay Details
| Gene Symbol: Fam120a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001033268.2 | NP_001028440.2 | 6 - 7 | 1126 | 104 | |
| GenBank mRNA | AK172909.1 | - | 6 - 7 | 1315 | 104 | |
| BC010304.1 | - | 2 - 3 | 194 | 104 | ||
| BC042582.1 | - | 5 - 6 | 642 | 104 | ||
| BC158134.1 | - | 6 - 7 | 1118 | 104 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Fam120a
Genomic Map