Assay Details
| Gene Symbol: Fam120a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001033268.2 | NP_001028440.2 | 7 - 8 | 1413 | 94 | |
| GenBank mRNA | AK172909.1 | - | 7 - 8 | 1602 | 94 | |
| AK219481.1 | - | 1 - 2 | 79 | 94 | ||
| BC010304.1 | - | 3 - 4 | 481 | 94 | ||
| BC042582.1 | - | 6 - 7 | 929 | 94 | ||
| BC158134.1 | - | 7 - 8 | 1405 | 94 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Fam120a
Genomic Map