Assay Details
| Gene Symbol: Fam120a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001033268.2 | NP_001028440.2 | 8 - 9 | 1497 | 97 | |
| GenBank mRNA | AK172909.1 | - | 8 - 9 | 1686 | 97 | |
| BC010304.1 | - | 4 - 5 | 565 | 97 | ||
| BC042582.1 | - | 7 - 8 | 1013 | 97 | ||
| BC158134.1 | - | 8 - 9 | 1489 | 97 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Fam120a
Genomic Map