Assay Details
| Gene Symbol: Fam120a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001033268.2 | NP_001028440.2 | 9 - 10 | 1726 | 102 | |
| GenBank mRNA | AK172909.1 | - | 9 - 10 | 1915 | 102 | |
| BC010304.1 | - | 5 - 6 | 794 | 102 | ||
| BC033412.1 | - | 1 - 2 | 54 | 102 | ||
| BC042582.1 | - | 8 - 9 | 1242 | 102 | ||
| BC158134.1 | - | 9 - 10 | 1718 | 102 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Fam120a
Genomic Map