Assay Details
| Gene Symbol: Crat | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_007760.3 | NP_031786.2 | 9 - 10 | 1614 | 73 | |
| GenBank mRNA | AK028666.1 | - | 3 - 4 | 259 | 73 | |
| AK041431.1 | - | 9 - 10 | 1389 | 73 | ||
| AK132179.1 | - | 10 - 11 | 1441 | 73 | ||
| AK170740.1 | - | 9 - 10 | 1400 | 73 | ||
| BC006668.1 | - | 9 - 10 | 1361 | 3499169 | 73 | |
| X85983.1 | - | 9 - 10 | 1332 | 73 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Crat
Genomic Map