Assay Details
| Gene Symbol: Gpr155 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001190297.2 | NP_001177226.1 | 10 - 11 | 1972 | 68 | |
| NM_001276443.1 | NP_001263372.1 | 10 - 11 | 1972 | 68 | ||
| NM_001276444.1 | NP_001263373.1 | 9 - 10 | 1888 | 68 | ||
| GenBank mRNA | AK089436.1 | - | 10 - 11 | 1890 | 68 | |
| AK165887.1 | - | 5 - 6 | 442 | 68 | ||
| AY255560.1 | - | 1 - 2 | 88 | 68 | ||
| BC115698.1 | - | 10 - 11 | 1868 | 40060105 | 68 | |
| HM161763.2 | - | 10 - 11 | 1949 | 68 | ||
| HM161764.2 | - | 10 - 11 | 1949 | 68 | ||
| HM161765.2 | - | 10 - 11 | 1949 | 68 | ||
| HM161766.2 | - | 9 - 10 | 1865 | 68 | ||
| KF358283.1 | - | 10 - 11 | 1949 | 68 | ||
| KF358284.1 | - | 9 - 10 | 1831 | 68 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Gpr155
Genomic Map