Assay Details
| Gene Symbol: Gpr155 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001190297.2 | NP_001177226.1 | 11 - 12 | 2175 | 69 | |
| NM_001276443.1 | NP_001263372.1 | 11 - 12 | 2175 | 69 | ||
| NM_001276444.1 | NP_001263373.1 | 10 - 11 | 2091 | 69 | ||
| GenBank mRNA | AK089436.1 | - | 11 - 12 | 2093 | 69 | |
| AY255560.1 | - | 2 - 3 | 291 | 69 | ||
| BC026382.1 | - | 1 - 2 | 191 | 69 | ||
| BC115698.1 | - | 11 - 12 | 2071 | 40060105 | 69 | |
| HM161763.2 | - | 11 - 12 | 2152 | 69 | ||
| HM161764.2 | - | 11 - 12 | 2152 | 69 | ||
| HM161765.2 | - | 11 - 12 | 2152 | 69 | ||
| HM161766.2 | - | 10 - 11 | 2068 | 69 | ||
| KF358283.1 | - | 11 - 12 | 2152 | 69 | ||
| KF358284.1 | - | 10 - 11 | 2034 | 69 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Gpr155
Genomic Map