Assay Details
| Gene Symbol: Gpr155 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001190297.2 | NP_001177226.1 | 13 - 14 | 2416 | 79 | |
| NM_001276443.1 | NP_001263372.1 | 13 - 14 | 2416 | 79 | ||
| NM_001276444.1 | NP_001263373.1 | 12 - 13 | 2332 | 79 | ||
| GenBank mRNA | AK089436.1 | - | 13 - 14 | 2334 | 79 | |
| BC026382.1 | - | 3 - 4 | 432 | 79 | ||
| BC115698.1 | - | 13 - 14 | 2312 | 40060105 | 79 | |
| HM161763.2 | - | 13 - 14 | 2393 | 79 | ||
| HM161764.2 | - | 13 - 14 | 2393 | 79 | ||
| HM161765.2 | - | 13 - 14 | 2393 | 79 | ||
| HM161766.2 | - | 12 - 13 | 2309 | 79 | ||
| KF358283.1 | - | 13 - 14 | 2393 | 79 | ||
| KF358284.1 | - | 12 - 13 | 2275 | 79 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Gpr155
Genomic Map