Assay Details
| Gene Symbol: Gpr155 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001190297.2 | NP_001177226.1 | 14 - 15 | 2515 | 85 | |
| NM_001276443.1 | NP_001263372.1 | 14 - 15 | 2515 | 85 | ||
| NM_001276444.1 | NP_001263373.1 | 13 - 14 | 2431 | 85 | ||
| GenBank mRNA | BC026382.1 | - | 4 - 5 | 531 | 85 | |
| HM161763.2 | - | 14 - 15 | 2492 | 85 | ||
| HM161764.2 | - | 14 - 15 | 2492 | 85 | ||
| HM161766.2 | - | 13 - 14 | 2408 | 85 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Gpr155
Genomic Map