Assay Details
| Gene Symbol: Gpr155 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001190297.2 | NP_001177226.1 | 3 - 4 | 868 | 131 | |
| NM_001276443.1 | NP_001263372.1 | 3 - 4 | 868 | 131 | ||
| NM_001276444.1 | NP_001263373.1 | 3 - 4 | 868 | 131 | ||
| GenBank mRNA | AK089436.1 | - | 3 - 4 | 786 | 131 | |
| BC115698.1 | - | 3 - 4 | 764 | 40060105 | 131 | |
| HM161763.2 | - | 3 - 4 | 845 | 131 | ||
| HM161764.2 | - | 3 - 4 | 845 | 131 | ||
| HM161765.2 | - | 3 - 4 | 845 | 131 | ||
| HM161766.2 | - | 3 - 4 | 845 | 131 | ||
| KF358283.1 | - | 3 - 4 | 845 | 131 | ||
| KF358284.1 | - | 3 - 4 | 845 | 131 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Gpr155
Genomic Map