Assay Details
| Gene Symbol: Slc27a4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011989.4 | NP_036119.1 | 11 - 12 | 1914 | 69 | |
| GenBank mRNA | AF072759.1 | - | 9 - 10 | 1216 | 69 | |
| AJ251113.1 | - | 10 - 11 | 1627 | 69 | ||
| AK009292.1 | - | 5 - 6 | 701 | 69 | ||
| AK033519.1 | - | 7 - 8 | 858 | 69 | ||
| AK036919.1 | - | 11 - 12 | 1916 | 69 | ||
| AK155313.1 | - | 11 - 12 | 1883 | 69 | ||
| AK155388.1 | - | 11 - 12 | 1884 | 69 | ||
| BC023114.1 | - | 11 - 12 | 1765 | 4158956 | 69 | |
| CT010312.1 | - | 10 - 11 | 1627 | 69 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Slc27a4
Genomic Map