Assay Details
| Gene Symbol: Sh3glb2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001289710.1 | NP_001276639.1 | 8 - 9 | 883 | 110 | |
| NM_139302.2 | NP_647463.1 | 9 - 10 | 946 | 110 | ||
| GenBank mRNA | AF426314.1 | - | 9 - 10 | 843 | 110 | |
| AK129455.1 | - | 8 - 9 | 2324 | 110 | ||
| AK152409.1 | - | 9 - 10 | 876 | 110 | ||
| AK152502.1 | - | 8 - 9 | 834 | 110 | ||
| AK167762.1 | - | 9 - 10 | 919 | 110 | ||
| BC024477.1 | - | 9 - 10 | 861 | 4977001 | 110 | |
| EF676083.1 | - | 9 - 10 | 922 | 110 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Sh3glb2
Genomic Map