Assay Details
| Gene Symbol: Sh3glb2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001289709.1 | NP_001276638.1 | 1 | 77 | 101 | |
| NM_001289710.1 | NP_001276639.1 | 1 | 77 | 101 | ||
| NM_139302.2 | NP_647463.1 | 1 | 77 | 101 | ||
| GenBank mRNA | AK167762.1 | - | 1 | 50 | 101 | |
| BC028859.1 | - | 1 | 66 | 4167974 | 101 | |
| BY317126.1 | - | 1 | 78 | 101 | ||
| CX216029.1 | - | 1 | 67 | 101 | ||
| EF676083.1 | - | 1 | 53 | 101 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK167762.1, BC028859.1, EF676083.1, NM_001289709.1, NM_001289710.1, NM_139302.2 transcripts.
Set Membership: |
|

Sh3glb2
Genomic Map