Assay Details
| Gene Symbol: Zfp786 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_177882.4 | NP_808550.1 | 2 - 3 | 243 | 94 | |
| GenBank mRNA | AK080438.1 | - | 2 - 3 | 243 | 94 | |
| BC138897.1 | - | 2 - 3 | 184 | 8861919 | 94 | |
More Information
Updated Mapping Information
Set Membership: |
|

Zfp786
Genomic Map