Assay Details
| Gene Symbol: Krba1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_133922.3 | NP_598683.3 | 12 - 13 | 2031 | 90 | |
| GenBank mRNA | AB041543.1 | - | 10 - 11 | 1663 | 90 | |
| AK029235.1 | - | 3 - 4 | 477 | 90 | ||
| AK040525.1 | - | 12 - 13 | 2033 | 90 | ||
| AK128931.1 | - | 12 - 13 | 1982 | 90 | ||
| AK162934.1 | - | 12 - 13 | 1739 | 90 | ||
| AK173281.1 | - | 12 - 13 | 3735 | 90 | ||
| BC066816.1 | - | 12 - 13 | 1951 | 30084911 | 90 | |
More Information
Set Membership: |
|

Krba1
Genomic Map