Assay Details
| Gene Symbol: Atp6v0e2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_133764.3 | NP_598525.1 | 2 - 3 | 337 | 61 | |
| GenBank mRNA | AK002260.1 | - | 2 - 3 | 205 | 61 | |
| AK077313.1 | - | 2 - 3 | 209 | 61 | ||
| BI134693.1 | - | 2 - 3 | 61 | |||
More Information
Set Membership: |
|

Atp6v0e2
Genomic Map