Assay Details
| Gene Symbol: Pax8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011040.4 | NP_035170.1 | 2 - 3 | 225 | 62 | |
| GenBank mRNA | AK050717.1 | - | 2 - 3 | 201 | 62 | |
| AK052610.1 | - | 2 - 3 | 201 | 62 | ||
| BC020526.1 | - | 2 - 3 | 192 | 4239835 | 62 | |
| X57487.1 | - | 2 - 3 | 182 | 62 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the BC020526.1, NM_011040.4, X57487.1 transcripts.
Set Membership: |
|

Pax8
Genomic Map