Assay Details
| Gene Symbol: Prr5l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001110849.1 | NP_001104319.1 | 2 - 3 | 157 | 126 | |
| GenBank mRNA | CJ073028.1 | - | 2 - 3 | 158 | 126 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001110849.1 transcripts.
Set Membership: |
|

Prr5l
Genomic Map