Assay Details
| Gene Symbol: Rapgef4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001204165.1 | NP_001191094.1 | 23 - 24 | 2326 | 59 | |
| NM_001204166.1 | NP_001191095.1 | 20 - 21 | 2218 | 59 | ||
| NM_019688.2 | NP_062662.1 | 22 - 23 | 2272 | 59 | ||
| GenBank mRNA | AB021132.1 | - | 23 - 24 | 2301 | 59 | |
| AB037668.1 | - | 14 - 15 | 1661 | 59 | ||
| AB445011.1 | - | 20 - 21 | 2210 | 59 | ||
| AF115480.1 | - | 22 - 23 | 2200 | 59 | ||
| AK004874.1 | - | 14 - 15 | 1318 | 59 | ||
| AK220281.1 | - | 23 - 24 | 2326 | 59 | ||
| BC139822.1 | - | 23 - 24 | 2258 | 40141811 | 59 | |
More Information
Updated Mapping Information
Set Membership: |
|

Rapgef4
Genomic Map