| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0200008555.mm.1 |
NM_001204167.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200008555.mm.1 |
NM_001204167.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200008555.mm.1 |
TC0200001049.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0200008557.mm.1 |
NM_001204167.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200008557.mm.1 |
NM_001204167.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200008557.mm.1 |
TC0200001049.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0200008558.mm.1 |
NM_001204167.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200008558.mm.1 |
NM_001204167.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200008558.mm.1 |
TC0200001049.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0200008553.mm.1 |
NM_001204167.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200008553.mm.1 |
NM_001204167.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200008553.mm.1 |
TC0200001049.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0200008556.mm.1 |
NM_001204167.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200008556.mm.1 |
NM_001204167.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200008556.mm.1 |
TC0200001049.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0200008554.mm.1 |
NM_001204167.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200008554.mm.1 |
NM_001204167.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200008554.mm.1 |
TC0200001049.mm.1 |
MTA TC |