Assay Details
| Gene Symbol: Fmn1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001285458.1 | NP_001272387.1 | 7 - 8 | 3244 | 108 | |
| NM_001285459.1 | NP_001272388.1 | 7 - 8 | 2868 | 108 | ||
| NM_010230.2 | NP_034360.2 | 9 - 10 | 3538 | 108 | ||
| GenBank mRNA | BC138036.1 | - | 6 - 7 | 3150 | 8861055 | 108 |
| BC171945.1 | - | 6 - 7 | 3144 | 9054231 | 108 | |
| CK391697.1 | - | 3 - 4 | 170 | 108 | ||
| CO814739.2 | - | 2 - 3 | 95 | 108 | ||
| X53599.1 | - | 9 - 10 | 3498 | 108 | ||
| X62379.1 | - | 7 - 8 | 2874 | 108 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Fmn1
Genomic Map