Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0200040308.mm.1 |
NM_029148.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200040308.mm.1 |
NM_029148.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200040308.mm.1 |
TC0200004691.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200040311.mm.1 |
NM_029148.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200040311.mm.1 |
NM_029148.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200040311.mm.1 |
TC0200004691.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200040313.mm.1 |
NM_029148.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200040313.mm.1 |
NM_029148.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200040313.mm.1 |
TC0200004691.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200040310.mm.1 |
NM_029148.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200040310.mm.1 |
NM_029148.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200040310.mm.1 |
TC0200004691.mm.1 |
MTA TC |