Assay Details
| Gene Symbol: Wars | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001164314.1 | NP_001157786.1 | 2 - 3 | 274 | 95 | |
| NM_001164488.1 | NP_001157960.1 | 2 - 3 | 207 | 95 | ||
| NM_011710.3 | NP_035840.3 | 2 - 3 | 220 | 95 | ||
| GenBank mRNA | AK004541.1 | - | 2 - 3 | 208 | 95 | |
| AK150090.1 | - | 2 - 3 | 238 | 95 | ||
| AK152966.1 | - | 2 - 3 | 238 | 95 | ||
| AK153337.1 | - | 2 - 3 | 246 | 95 | ||
| BC046232.1 | - | 2 - 3 | 213 | 6813406 | 95 | |
| BY773669.1 | - | 2 - 3 | 275 | 95 | ||
| CD563585.1 | - | 2 - 3 | 194 | 95 | ||
More Information
Set Membership: |
|

Wars
Genomic Map