Assay Details
| Gene Symbol: Kif16b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081133.2 | NP_001074602.1 | 10 - 11 | 1268 | 125 | |
| GenBank mRNA | AB610968.1 | - | 10 - 11 | 1323 | 125 | |
| AK033018.1 | - | 4 - 5 | 595 | 125 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Kif16b
Genomic Map