Assay Details
| Gene Symbol: Yy1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009537.3 | NP_033563.2 | 3 - 4 | 1007 | 125 | |
| GenBank mRNA | AK076149.1 | - | 4 - 5 | 620 | 125 | |
| AK166542.1 | - | 4 - 5 | 345 | 125 | ||
| BC055899.1 | - | 3 - 4 | 985 | 3156776 | 125 | |
| M73963.1 | - | 3 - 4 | 1007 | 125 | ||
| M74590.1 | - | 3 - 4 | 947 | 125 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Yy1
Genomic Map