Assay Details
| Gene Symbol: Csrnp3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_153409.5 | NP_700458.3 | 2 - 3 | 269 | 119 | |
| GenBank mRNA | AK020730.1 | - | 2 - 3 | 192 | 119 | |
| AK039150.1 | - | 2 - 3 | 195 | 119 | ||
| AK080761.1 | - | 2 - 3 | 192 | 119 | ||
| BY297754.1 | - | 2 - 3 | 271 | 119 | ||
| EF210820.1 | - | 2 - 3 | 241 | 119 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK039150.1, EF210820.1, NM_153409.5 transcripts.
Set Membership: |
|

Csrnp3
Genomic Map