Assay Details
| Gene Symbol: Dgkz | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001166597.1 | NP_001160069.1 | 24 - 25 | 2883 | 73 | |
| NM_138306.2 | NP_612179.2 | 23 - 24 | 2282 | 73 | ||
| GenBank mRNA | AK087503.1 | - | 24 - 25 | 73 | ||
| AK170936.1 | - | 20 - 21 | 1905 | 73 | ||
| AK204403.1 | - | 3 - 4 | 211 | 73 | ||
| BC014860.1 | - | 8 - 9 | 752 | 73 | ||
| BC049228.1 | - | 23 - 24 | 2318 | 3668704 | 73 | |
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Transcript Location | Amplicon Length | |
|---|---|---|---|---|---|---|---|
| GenBankmRNA | AK088841.1 | - | Chr. - - [-] on Build | 73 | |||
More Information
Updated Mapping Information
Set Membership: |
|

Dgkz
Genomic Map