Assay Details
| Gene Symbol: Dgkz | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001166597.1 | NP_001160069.1 | 29 - 30 | 3258 | 70 | |
| NM_138306.2 | NP_612179.2 | 28 - 29 | 2657 | 70 | ||
| GenBank mRNA | AK087503.1 | - | 29 - 30 | 3260 | 70 | |
| AK170936.1 | - | 25 - 26 | 2280 | 70 | ||
| BC014860.1 | - | 13 - 14 | 1127 | 70 | ||
| BC049228.1 | - | 28 - 29 | 2693 | 3668704 | 70 | |
| BE862117.1 | - | 5 - 6 | 386 | 70 | ||
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Transcript Location | Amplicon Length | |
|---|---|---|---|---|---|---|---|
| GenBankmRNA | AK088841.1 | - | Chr. - - [-] on Build | 70 | |||
More Information
Updated Mapping Information
Set Membership: |
|

Dgkz
Genomic Map