Assay Details
| Gene Symbol: Fubp3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001033389.4 | NP_001028561.2 | 3 - 4 | 449 | 109 | |
| NM_001290548.1 | NP_001277477.1 | 3 - 4 | 449 | 109 | ||
| GenBank mRNA | AK145377.1 | - | 3 - 4 | 284 | 109 | |
| AK166081.1 | - | 3 - 4 | 292 | 109 | ||
| AK167671.1 | - | 3 - 4 | 269 | 109 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Fubp3
Genomic Map