Assay Details
| Gene Symbol: Fubp3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001033389.4 | NP_001028561.2 | 6 - 7 | 626 | 78 | |
| NM_001290548.1 | NP_001277477.1 | 6 - 7 | 626 | 78 | ||
| GenBank mRNA | AK145377.1 | - | 6 - 7 | 461 | 78 | |
| AK166081.1 | - | 6 - 7 | 469 | 78 | ||
| AK166478.1 | - | 6 - 7 | 461 | 78 | ||
| AK167671.1 | - | 6 - 7 | 446 | 78 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Fubp3
Genomic Map