Assay Details
| Gene Symbol: Exosc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_144886.2 | NP_659135.1 | 1 - 2 | 127 | 76 | |
| GenBank mRNA | AK144234.1 | - | 1 - 2 | 136 | 76 | |
| AK164618.1 | - | 1 - 2 | 146 | 76 | ||
| AK166881.1 | - | 1 - 2 | 137 | 76 | ||
| BC021485.1 | - | 1 - 2 | 151 | 5344620 | 76 | |
| BC021807.1 | - | 1 - 2 | 149 | 3980155 | 76 | |
More Information
Updated Mapping Information
Set Membership: |
|

Exosc2
Genomic Map