Assay Details
| Gene Symbol: Exosc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_144886.2 | NP_659135.1 | 3 - 4 | 266 | 68 | |
| GenBank mRNA | AK144234.1 | - | 3 - 4 | 275 | 68 | |
| AK164618.1 | - | 3 - 4 | 285 | 68 | ||
| AK166881.1 | - | 3 - 4 | 276 | 68 | ||
| BC021485.1 | - | 3 - 4 | 290 | 5344620 | 68 | |
| BC021807.1 | - | 3 - 4 | 288 | 3980155 | 68 | |
More Information
Set Membership: |
|

Exosc2
Genomic Map