Assay Details
| Gene Symbol: Exosc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_144886.2 | NP_659135.1 | 7 - 8 | 670 | 90 | |
| GenBank mRNA | AK144234.1 | - | 7 - 8 | 679 | 90 | |
| AK164618.1 | - | 6 - 7 | 1452 | 90 | ||
| AK166881.1 | - | 7 - 8 | 680 | 90 | ||
| BC021485.1 | - | 7 - 8 | 694 | 5344620 | 90 | |
| BC021807.1 | - | 7 - 8 | 692 | 3980155 | 90 | |
More Information
Updated Mapping Information
Set Membership: |
|

Exosc2
Genomic Map