Assay Details
| Gene Symbol: Fbxo3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_020593.2 | NP_065618.1 | 5 - 6 | 740 | 73 | |
| NM_212433.1 | NP_997598.1 | 5 - 6 | 740 | 73 | ||
| GenBank mRNA | AF233226.1 | - | 5 - 6 | 679 | 73 | |
| AK004544.2 | - | 5 - 6 | 692 | 73 | ||
| AK009024.1 | - | 5 - 6 | 679 | 73 | ||
| AK028048.1 | - | 5 - 6 | 702 | 73 | ||
| AK031185.1 | - | 5 - 6 | 679 | 73 | ||
| AK031428.1 | - | 5 - 6 | 700 | 73 | ||
| AK049870.1 | - | 5 - 6 | 698 | 73 | ||
| AK050195.1 | - | 5 - 6 | 692 | 73 | ||
| AK085686.1 | - | 5 - 6 | 695 | 73 | ||
| AK150097.1 | - | 5 - 6 | 692 | 73 | ||
| AK151130.1 | - | 5 - 6 | 691 | 73 | ||
| AK151857.1 | - | 5 - 6 | 691 | 73 | ||
| AK152780.1 | - | 5 - 6 | 691 | 73 | ||
| AK167075.1 | - | 5 - 6 | 678 | 73 | ||
| BC010212.1 | - | 5 - 6 | 641 | 73 | ||
| BC050758.1 | - | 5 - 6 | 693 | 73 | ||
| BC058253.1 | - | 5 - 6 | 697 | 6465107 | 73 | |
More Information
Updated Mapping Information
Set Membership: |
|

Fbxo3
Genomic Map