Assay Details
| Gene Symbol: Fbxo3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_020593.2 | NP_065618.1 | 6 - 7 | 789 | 93 | |
| NM_212433.1 | NP_997598.1 | 6 - 7 | 789 | 93 | ||
| GenBank mRNA | AF233226.1 | - | 6 - 7 | 728 | 93 | |
| AK004544.2 | - | 6 - 7 | 741 | 93 | ||
| AK009024.1 | - | 6 - 7 | 728 | 93 | ||
| AK019550.1 | - | 6 - 7 | 729 | 93 | ||
| AK028048.1 | - | 6 - 7 | 751 | 93 | ||
| AK031185.1 | - | 6 - 7 | 728 | 93 | ||
| AK031428.1 | - | 6 - 7 | 749 | 93 | ||
| AK049870.1 | - | 6 - 7 | 747 | 93 | ||
| AK050195.1 | - | 6 - 7 | 741 | 93 | ||
| AK085686.1 | - | 6 - 7 | 744 | 93 | ||
| AK150097.1 | - | 6 - 7 | 741 | 93 | ||
| AK152780.1 | - | 6 - 7 | 740 | 93 | ||
| AK167075.1 | - | 6 - 7 | 727 | 93 | ||
| BC010212.1 | - | 6 - 7 | 690 | 93 | ||
| BC050758.1 | - | 6 - 7 | 742 | 93 | ||
| BC058253.1 | - | 6 - 7 | 746 | 6465107 | 93 | |
More Information
Updated Mapping Information
Set Membership: |
|

Fbxo3
Genomic Map