Assay Details
| Gene Symbol: Sel1l2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001033296.2 | NP_001028468.1 | 11 - 12 | 1107 | 74 | |
| GenBank mRNA | AK016809.1 | - | 11 - 12 | 1053 | 74 | |
| AK132650.1 | - | 11 - 12 | 1108 | 74 | ||
| BC150948.1 | - | 11 - 12 | 1079 | 9087859 | 74 | |
More Information
Updated Mapping Information
Set Membership: |
|

Sel1l2
Genomic Map