Assay Details
| Gene Symbol: Rabl6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001024616.1 | NP_001019787.2 | 10 - 11 | 1523 | 72 | |
| GenBank mRNA | AK045520.1 | - | 10 - 11 | 1545 | 72 | |
| AK081440.1 | - | 10 - 11 | 1533 | 72 | ||
| AK157020.1 | - | 10 - 11 | 1558 | 72 | ||
| AK162461.1 | - | 6 - 7 | 903 | 72 | ||
| AK171643.1 | - | 10 - 11 | 1534 | 72 | ||
| AK215518.1 | - | 2 - 3 | 296 | 72 | ||
| AK219937.1 | - | 2 - 3 | 350 | 72 | ||
| BC006855.1 | - | 2 - 3 | 3255 | 72 | ||
| BC020111.1 | - | 10 - 11 | 1531 | 72 | ||
| BC021476.1 | - | 10 - 11 | 1538 | 72 | ||
| BC023692.1 | - | 10 - 11 | 1551 | 72 | ||
| BC040791.1 | - | 2 - 3 | 3255 | 72 | ||
| BC051138.1 | - | 10 - 11 | 1537 | 72 | ||
| BC058585.1 | - | 10 - 11 | 1536 | 72 | ||
| BC085507.1 | - | 10 - 11 | 1523 | 5716088 | 72 | |
More Information
Updated Mapping Information
Set Membership: |
|

Rabl6
Genomic Map