Assay Details
| Gene Symbol: Rabl6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001024616.1 | NP_001019787.2 | 2 - 3 | 464 | 81 | |
| GenBank mRNA | AK045520.1 | - | 2 - 3 | 486 | 81 | |
| AK081440.1 | - | 2 - 3 | 474 | 81 | ||
| AK143678.1 | - | 2 - 3 | 502 | 81 | ||
| AK157020.1 | - | 2 - 3 | 500 | 81 | ||
| AK171643.1 | - | 2 - 3 | 475 | 81 | ||
| AK196492.1 | - | 1 - 2 | 116 | 81 | ||
| BC020111.1 | - | 2 - 3 | 472 | 81 | ||
| BC021476.1 | - | 2 - 3 | 479 | 81 | ||
| BC023692.1 | - | 2 - 3 | 492 | 81 | ||
| BC051138.1 | - | 2 - 3 | 483 | 81 | ||
| BC058585.1 | - | 2 - 3 | 477 | 81 | ||
| BC085507.1 | - | 2 - 3 | 464 | 5716088 | 81 | |
More Information
Updated Mapping Information
Set Membership: |
|

Rabl6
Genomic Map