| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0200026198.mm.1 |
NM_001024616.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200026198.mm.1 |
NM_001024616.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200026198.mm.1 |
TC0200003162.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0200026200.mm.1 |
NM_001024616.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200026200.mm.1 |
NM_001024616.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200026200.mm.1 |
TC0200003162.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0200026205.mm.1 |
NM_001024616.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200026205.mm.1 |
NM_001024616.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200026205.mm.1 |
TC0200003162.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0200026197.mm.1 |
NM_001024616.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200026197.mm.1 |
NM_001024616.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200026197.mm.1 |
TC0200003162.mm.1 |
MTA TC |